Impurity and API standard, 5-(1-methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol hydrochloride
PROOMM0609.07
:
- Pk:50 mg
- Standards:Reference standards
- CAS No.:4046-24-6
- Description:5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol hydrochloride
- Formula:C₂₁H₂₄ClNO
- Description:5-(1-Methylpiperidin-4-yl)-5H-dibenzo[a,d][7]annulen-5-ol hydrochloride