ContactUsLinkComponent
VWR®, Workstation Dispenser
VWR®, Workstation Dispenser
Codice catalogo 216-2006
Nome del fornitore:  VWR Collection
undefined
VWR®, Workstation Dispenser
Codice catalogo 216-2006
Nome del fornitore:  VWR Collection
Specifiche
  • Colour:
    Clear
  • Description:
    Workstation dispenser
  • W×D×H:
    265×137×440 mm
  • Pk:
    1
Specifiche
Informazioni su questo articolo
This acrylic combination accessory dispenser holds the most commonly used laboratory disposables in one central location. Dispenser stores tubes, tips, gloves, Kimwipes® and Parafilm®.
  • Two compartments with hinged lid
  • One compartment for wipe boxes and one for tape
  • One large compartment for gloves or wipe boxes