Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, 16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
Impurity and API standard, 16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
Catalog # PROOMM0204.02
CAS Number:  
Impurity and API standard, 16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
Catalog # PROOMM0204.02
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    1040085-98-0
  • Description:
    16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione
  • Formula:
    C₂₃H₃₀O₆
  • Description:
    16alpha,17-[(1RS)-Ethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione

Specifications

About this item