Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol
Impurity and API standard, (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol
Catalog # PROOMM0180.08
CAS Number:  
Impurity and API standard, (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol
Catalog # PROOMM0180.08
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • CAS No.:
    59743-88-3
  • Description:
    (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol
  • Formula:
    C₂₀H₂₃NO₂S
  • Description:
    (4RS)-10-Methoxy-4-(1-methylpiperidin-4-yl)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ol

Specifications

About this item