Order Entry
Switzerland
ContactUsLinkComponent
 
Impurity and API standard, [2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol
  PROOMM0168.04
 :  
Impurity and API standard, [2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol
  PROOMM0168.04
 :  

 

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    114799-13-2
  • Description:
    [2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol
  • Formula:
    C₂₂H₂₃ClN₆O
  • Description:
    [2-Butyl-5-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-4-yl]methanol