Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, (4RS)-3-amino-6-chloro-2-methyl-4-phenyl-3,4-dihydroquinazolin-4-ol
Impurity and API standard, (4RS)-3-amino-6-chloro-2-methyl-4-phenyl-3,4-dihydroquinazolin-4-ol
Catalog # PROOMM0136.05
CAS Number:  
Impurity and API standard, (4RS)-3-amino-6-chloro-2-methyl-4-phenyl-3,4-dihydroquinazolin-4-ol
Catalog # PROOMM0136.05
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    27537-87-7
  • Description:
    (4RS)-3-Amino-6-chloro-2-methyl-4-phenyl-3,4-dihydroquinazolin-4-ol
  • Formula:
    C₁₅H₁₄ClN₃O
  • Description:
    (4RS)-3-Amino-6-chloro-2-methyl-4-phenyl-3,4-dihydroquinazolin-4-ol

Specifications

About this item