Impurity and API standard, 7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one
Catalog # PROOMM0064.09
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:868350-97-4
- Description:7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one
- Formula:C₁₅H₁₂BrN₃O
- Description:7-Bromo-5-(6-methylpyridin-2-yl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one