Impurity and API standard, 2-Bromo-N-[4-bromo-2-(pyridin-2-ylcarbonyl)phenyl]acetamide
Catalog # PROOMM0064.07
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:1694-64-0
- Description:2-Bromo-N-[4-bromo-2-(pyridin-2-ylcarbonyl)phenyl]acetamide
- Formula:C₁₄H₁₀Br₂N₂O₂
- Description:2-Bromo-N-[4-bromo-2-(pyridin-2-ylcarbonyl)phenyl]acetamide