Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, 3-amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one
Impurity and API standard, 3-amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one
Catalog # PROOMM0064.04
CAS Number:  
Impurity and API standard, 3-amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one
Catalog # PROOMM0064.04
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    77616-97-8
  • Description:
    3-Amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one
  • Formula:
    C₁₄H₁₀BrN₃O
  • Description:
    3-Amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one

Specifications

About this item