Impurity and API standard, enalaprilat
PROOMM0010.01
:
- Pk:100 mg
- Standards:Reference standards
- CAS No.:76420-72-9
- Description:(2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid
- Formula:C₁₈H₂₄N₂O₅
- Description:(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid (Enalaprilat)