Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, Levomepromazine meleate assay standard, BP
Organic reference standards, Levomepromazine meleate assay standard, BP
Catalog # PROOBP723
CAS Number:  
Organic reference standards, Levomepromazine meleate assay standard, BP
Catalog # PROOBP723
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standard
  • CAS No.:
    60-99-1
  • Description:
    Levomepromazine meleate assay standard
  • Formula:
    C₁₉H₂₄N₂OS
  • Description:
    Levomepromazine maleate assay standard

Specifications

About this item