Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, Cloprostenol assay standard, BP
Organic reference standards, Cloprostenol assay standard, BP
Catalog # PROOBP628
CAS Number:  
Organic reference standards, Cloprostenol assay standard, BP
Catalog # PROOBP628
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    55028-72-3
  • Description:
    Cloprostenol assay standard
  • Formula:
    C₂₂H₂₈ClO₆·Na
  • Description:
    Cloprostenol sodium assay standard

Specifications

About this item