Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, hyoscine hydrobromide assay standard, BP
Organic reference standards, hyoscine hydrobromide assay standard, BP
Catalog # PROOBP199
CAS Number:  
Organic reference standards, hyoscine hydrobromide assay standard, BP
Catalog # PROOBP199
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Standards:
    Reference standard
  • CAS No.:
    114-49-8
  • Description:
    Hyoscine hydrobromide assay standard
  • Formula:
    C₁₇H₂₂BrNO₄
  • Description:
    Hyoscine hydrobromide assay standard

Specifications

About this item