Organic reference standards, glibenclamide assay standard, BP
Catalog # PROOBP175
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference standard
- CAS No.:10238-21-8
- Description:Glibenclamide assay standard
- Formula:C₂₃H₂₈ClN₃O₅S
- Description:Glibenclamide assay standard