ContactUsLinkComponent
Detergents, acid neutraliser, neodisher® Z
Detergents, acid neutraliser, neodisher® Z
Catalog # MIELUS483030
Supplier:  MIELE
Detergents, acid neutraliser, neodisher® Z
Catalog # MIELUS483030
Supplier:  MIELE
Specifications
  • Description:
    neodisher® Z acid neutraliser detergent
  • Packed:
    5 l canister
  • Pk:
    1
Specifications
About this item
neodisher® Z is a acid neutralising agent for the automated reprocessing of laboratory glassware for sensitive water examinations, laboratories in phosphate industry and also for the automated reprocessing of medical utensils in hospitals.
  • Free of surfactants and phosphates
  • Based on organic acids
  • Used to remove acid-soluble residues in laboratories where residues of phosphates, nitrogen and surfactants would adulterate sensitive analyses
neodisher® Z cleans stainless steel instruments, utensils and laboratory glassware as well as acidic pre-cleaning of laboratory glassware.
Certifications: Conforms to the European directive 93/42/EC, Annex I, concerning medical devices.
Caution: For avoiding water stains the use of deionised water in the final rinse is recommended.