Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, moxidectin, Ph. Eur. RS
Organic reference standards, moxidectin, Ph. Eur. RS
Supplier:  EDQM
Certificates
Organic reference standards, moxidectin, Ph. Eur. RS
Supplier:  EDQM

Specifications

  • CAS No.:
    113507-06-5
  • Formula:
    C₃₇H₅₃NO₈
  • Description:
    Moxidectin

Specifications

Frequently Bought Together