Water for dilution organic-free
Catalog # 705-2037
Supplier: Hach
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
About this item
Synonyms: Hydrogen oxide
:
:18.02 g/mol
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332