ContactUsLinkComponent
General purpose gloves, FineDex® 944-01 Clean
General purpose gloves, FineDex® 944-01 Clean
Supplier:  HONEYWELL SAFETY
Certificates
undefined
General purpose gloves, FineDex® 944-01 Clean
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALL2094401/6
  • DALL2094401/10
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Finish:
  • Diamond-pattern
  • Diamond-pattern
  • Cuff style:
  • Pinked
  • Pinked
  • EN norm:
  • EN 388:2016 (1010X), EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
  • EN 388:2016 (1010X), EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)