Order Entry
Switzerland
ContactUsLinkComponent
Rat TNF-α ELISA kit
Rat TNF-α ELISA kit
Catalog # BIRBORB79137-96
Supplier:  Biorbyt
CAS Number:  
Rat TNF-α ELISA kit
Catalog # BIRBORB79137-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Rat
  • Target protein:
    TNF-α
  • Description:
    Rat TNF-α ELISA Kit, 96 tests
  • Detection range:
    25 - 20000 pg/ml
  • Sensitivity:
    25 pg/ml

Specifications

About this item

Rat TNF-alpha ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat TNF-alpha in serum, plasma, cell culture supernatants and urine.

The assay employs an antibody specific for Rat TNF-alpha coated on a 96-well plate. Standards and samples are pipetted into the wells and TNF-alpha present in a sample is bound to the wells by the immobilised antibody. The wells are washed and biotinylated anti-Rat TNF-alpha antibody is added. After washing away unbound biotinylated antibody, HRP-conjugated streptavidin is pipetted to the wells. The wells are again washed, a TMB substrate solution is added to the wells and colour develops in proportion to the amount of TNF-alpha bound. The stop solution changes the colour from blue to yellow, and the intensity of the colour is measured at 450 nm.

Delivery: Kit contains pre-coated 96-well strip microplate, wash buffer, assay diluent(s), biotinylated detection antibody, HRP-streptavidin, TMB One-Step substrate, stop solution and lyophilised standard.