Order Entry
Switzerland
ContactUsLinkComponent
 
Test kits, photometric, Zirconium, LCK cuvette tests
Catalog # HACHLCK364
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, Zirconium, LCK cuvette tests
Catalog # HACHLCK364
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    SurTec/Hach Method
  • Pk:
    25 Tests
  • Description:
    Zirconium
  • Seq:
    296
  • Type:
    LCK364
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    6 - 60 mg/l

Specifications

About this item

The first specific, easy to use rapid test which helps to evaluate the Zirconium concentration during the passivation process.

  • Helps adjusting the right amount of Zirconium within the process
  • Helps minimising chemistry consumption and costs in Zirconium based coating processes
  • Helps ensuring a consistent high product quality

Storage conditions: 2 to 8 °C (keep refrigerated)