ContactUsLinkComponent
Test kits, photometric, Water hardness, residual, LCK cuvette tests
Test kits, photometric, Water hardness, residual, LCK cuvette tests
  705-3041
 :  Hach
 :  
Test kits, photometric, Water hardness, residual, LCK cuvette tests
  705-3041
 :  Hach
 :  
  • Chemical method:
    Metalphthalein
  • Pk:
    24 Tests
  • Description:
    Hardness (residual Ca + Mg)
  • Seq:
    235
  • Type:
    LCK427
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,02 - 0,6 °dH
    0,1 - 2 mg/l Ca
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C