Order Entry
Switzerland
ContactUsLinkComponent
Test kits, photometric, Surfactants, anionic, LCK cuvette tests
Test kits, photometric, Surfactants, anionic, LCK cuvette tests
Catalog # HACHLCK432
Supplier:  Hach
CAS Number:  
Test kits, photometric, Surfactants, anionic, LCK cuvette tests
Catalog # HACHLCK432
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Methylene Blue (MBA)
  • Pk:
    25 Tests
  • Description:
    Surfactants, anionic
  • Type:
    LCK432
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,1 - 4,0 mg/l

Specifications

About this item

The standard anionic surfactant method has several drawbacks:
*Multiple handling steps are time consuming and complicated.
*Large volumes of extraction solvents create a safety and environmental risk.
*The combination of lengthy handling time, glassware maintenance, and solvent disposal adds up to significant costs per test. A standardised cuvette-based extraction and photometric measurement system overcomes all these disadvantages to provide accurate, reproducible results.

  • Easy to use
  • Safe to handle
  • Cost saving
  • Providing accurate and reproducible results
  • Good phase separation even in samples with high organic load

Storage conditions: 15 to 25 °C

Certifications: ISO 7875-1-2-1984, DIN 38409-H 23-1