Test kits, photometric, Silver, LCK cuvette tests
Anbieter: Hach
Zertifikate
Über diesen Artikel
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C (LCK354)
Storage conditions: 2 to 8 °C (keep refrigerated) (LCK355)
Frequently Bought Together