Order Entry
Switzerland
ContactUsLinkComponent
Test kits, Photometric Iodine sample (PIS) according to MEBAK II standard, LCK cuvette tests
Test kits, Photometric Iodine sample (PIS) according to MEBAK II standard, LCK cuvette tests
Catalog # HACHLCK240
Supplier:  Hach
CAS Number:  
undefined
Test kits, Photometric Iodine sample (PIS) according to MEBAK II standard, LCK cuvette tests
Catalog # HACHLCK240
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    MEBAK Method
  • Pk:
    24 Tests
  • Description:
    Photometric iodine sample
  • Seq:
    263
  • Type:
    LCK240
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    > 0,2

Specifications

About this item

For Iodine values >0,2.

Storage conditions: 15 to 25 °C

Certifications: MEBAK II