Order Entry
Switzerland
ContactUsLinkComponent
Test kits, photometric, AOX, LCK cuvette tests
Test kits, photometric, AOX, LCK cuvette tests
Catalog # 705-3009
Supplier:  Hach
CAS Number:  
Test kits, photometric, AOX, LCK cuvette tests
Catalog # 705-3009
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Digestion + Iron(III)-Thiocyanate
  • Pk:
    24 Tests
  • Description:
    AOX
  • Seq:
    204
  • Type:
    LCK390.
  • For unit:
    DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
  • Range:
    0,05 - 3,0 mg/l AOX

Specifications

About this item

For the photometric determination of adsorbable organic halogens.

  • Easy to use Packaged in a fully equipped box

Storage conditions: 2 to 8 °C (keep refrigerated)

Certifications: DIN EN ISO 9562