Orders LinkContactUsLinkComponent
Laboratory blenders
Laboratory blenders
Supplier:   WARING BLENDOR
Laboratory blenders
Supplier:  WARING BLENDOR
Certificates
Laboratory blenders
Laboratory blenders
2 Options Available Below
Product Details & Documents
The CB15 series blenders are designed specifically for processing large quantities of free flowing material with a high liquid content. This range of 3-speed blenders has an integral pulse button and is fitted with a powerful 3,75 hp motor encased within a die-cast epoxy coated motor housing.
  • Easy to move and fitted with full traction rubber feet for greater stability
  • Easy-to-clean electronic keypad
  • Slow Start™ blending mechanism
  • Stainless steel container jar with two handles
  • Smaller 2, 1 litre and mini-containers can be used with the aid of the AD adapters
In addition the CB15VXE model has an a built in tachometer display showing the RPM of the motor. This makes it suitable for a range of validated applications such as GMP/GLP.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • WARICB15E
    Blender, 3 speed, container with PC see through lid, CB15E
    Blender, 3 speed, container with PC see through lid, CB15E
    4 l
    311,5×381×140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3’070.00
    SFr. 3’070.00
    1 * 1 ST
       SFr. 3’070.00
  • WARICB15TE
    Blender, 3 speed with timer, container with PC see through lid, CB15TE
    Blender, 3 speed with timer, container with PC see through lid, CB15TE
    4 l
    311,5×381×140 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 3’250.00
    SFr. 3’250.00
    1 * 1 ST
       SFr. 3’250.00