Order Entry
Switzerland
ContactUsLinkComponent
Acetic acid 1 mol/l (1 N), Titripur®, Supelco®
Acetic acid 1 mol/l (1 N), Titripur®, Supelco®
Catalog # 1.60305.1000
Supplier:  Merck
CAS Number:  
undefined
Acetic acid 1 mol/l (1 N), Titripur®, Supelco®
Catalog # 1.60305.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :60.05 g/mol
ADR:8,II
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152