Mca-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-Lys(Dnp)-Arg-Arg-NH₂ trifluoroacetate salt peptide
Supplier: BACHEM BIOCHEMICA
Certificates
About this item
This fluorescent (FRET) peptide substrate contains the 'Swedish' Lys-Met/Asn-Leu mutation of the amyloid precursor protein (APP) β-secretase cleavage site. Mca-SEVNLDAEFRK(Dnp)RR-amide has been used for assaying β-secretase-like activity of thimet oligopeptidase.
- Mca-(Asn⁶⁷⁰,Leu⁶⁷¹)-Amyloid β/A4 Protein Precursor₇₇₀ (667-676)-Lys(Dnp)-Arg-Arg amide trifluoroacetate salt
- Mca-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-Lys(Dnp)-Arg-Arg-NH₂ trifluoroacetate salt
Specifications
- CAS No.:438625-61-7
- Storage conditions:-20±5 °C
- Protein/peptide name:Mca-Ser-Glu-Val-Asn-Leu-Asp-Ala-Glu-Phe-Arg-Lys(Dnp)-Arg-Arg-NH₂ Trifluoroacetate salt Peptide
- Molecular formula:C₈₆H₁₂₅N₂₇O₂₉
Specifications
Frequently Bought Together