ContactUsLinkComponent
Test kits, total hardness determination by titration methods
Test kits, total hardness determination by titration methods
Catalog # HACH2447600
Supplier:  Hach
CAS Number:  
undefined
Test kits, total hardness determination by titration methods
Catalog # HACH2447600
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Hardness (Total) reagent set, low range
  • Seq:
    01
  • Range:
    0 - 2500 mg/l
Specifications
About this item
The kits are used to determine the total hardness level.
Delivery: Kit includes buffer solution (Hardness 1), ManVer® 2 hardness indicator powder pillows and TitraVer® (EDTA) and relevant titrant solution.