Order Entry
Switzerland
ContactUsLinkComponent
Urea 99.0-100.5%, BAKER ANALYZED® ACS, J.T.Baker®
Irritant
Health
Urea 99.0-100.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Health
Urea 99.0-100.5%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 100.5%
  • For Laboratory,Research,or Manufacturing Use:
    132 - 135 °C
  • Assay:
    99.0 - 100.5 %
  • Melting Point:
    132 - 135 °C
  • Insoluble Matter:
    ≤0.01 %
  • Residue after Ignition:
    ≤0.010 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Chloride (Cl):
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together