Order Entry
Switzerland
ContactUsLinkComponent
Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Health
Irritant
Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9450-R
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Tetrahydrofuran ≥99.5% (by GC, corrected for water content) stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 9450-R
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    400 lb
  • Seq:
    7284

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay (C₄H₈O) (by GC, corrected for water):
    ≥99.5 %
  • Color (APHA):
    ≤20
  • Peroxide (as H₂O₂):
    ≤0.015 %
  • Residue after Evaporation:
    ≤0.0300 %
  • Water (by KF, coulometric):
    ≤0.05 %
  • Specific Gravity at 25°/25°C:
    0.884 - 0.886
  • Stabilizer (BHT):
    100 - 400 ppm
  • Acidity:
    Passes Test

Specification Test Results

About this item

 :
 :72.11 g/mol
Merck Index:12,09351
Boiling Pt:66 °C (1013 hPa)
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
ADR:3,II
 :Management of Change (MOC) category = N
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 - 400 ppm
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356