Order Entry
Switzerland
ContactUsLinkComponent
Tin(II) chloride dihydrate 98.0-103%, crystals, AR® ACS, Macron Fine Chemicals™
Corrosive
Irritant
Tin(II) chloride dihydrate 98.0-103%, crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 8176-20
CAS Number:  
undefined
Corrosive
Irritant
Tin(II) chloride dihydrate 98.0-103%, crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 8176-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    98.0 - 103.0 %
  • ACS - Assay (SnCl₂·2H₂O):
    98.0 - 103.0 %
  • ACS - Mercury (Hg):
    ≤0.050 ppm
  • ACS - Solubility in HCl:
    Passes Test
  • ACS - Sodium (Na):
    ≤0.01 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.003 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Lead (Pb):
    ≤0.01 %

Specification Test Results

About this item

 :
 :225.65 g/mol
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:10025-69-1
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863