
Sodium chloride 99.0-100.5% (dried basis), granular USP, FCC, J.T.Baker®
Catalog # 3301-37
Supplier: Avantor
CAS Number:
Specifications
- Pk:12 kg
- Seq:4187
Specifications
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity or Alkalinity:Passes Test
- USP - Appearance of Solution:Passes Test
- USP - Aluminum (Al):≤ 0.2 ppm
- USP - Arsenic (As):≤ 1 ppm
- USP - Barium (Ba):Passes Test
- USP - Bromide (Br):≤ 100 ppm
- USP - Ferrocyanide:Passes Test
- USP - Iodides (I):Passes Test
- USP - Iron (Fe):≤ 2 ppm
- USP - Loss on Drying at 105°C:≤ 0.5 %
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤ 100 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤ 0.01
- USP - Phosphate (PO₄):≤ 25 ppm
- USP - Potassium (K):≤ 500 ppm
- USP - Sulfate (SO₄):≤ 200 ppm
- USP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- FCC - Assay (as NaCl) (FCC) (ignited basis):99.0 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Arsenic (As):≤ 1 ppm
- FCC - Calcium and Magnesium:≤ 0.35 %
- FCC - Heavy Metals (as Pb):≤ 2 ppm
- FCC - Loss on Drying at 105°C:≤ 0.5 %
- Solution Test:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:≤1 ppm
- manufacturing or purification of the product.:≤0.35%
- Must be subjected to further processing during the preparation of:≤2 ppm
- injectable dosage forms:≤0.5%
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:Passes Test
Specification Test Results
About this item
:
:58.44 g/mol
Melting Pt:801 °C
Density:2.16 g/ml (25 °C)
:Management of Change (MOC) category = HR
:GHS07
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477