
Sodium bromide ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7647-15-6
:
Formula:NaBr
MDL Number:MFCD00003475
:102.89 g/mol
Merck Index:13,08666
Melting Pt:753 °C
Density:3.21 g/cm³
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:≤0.005%
- ACS - Assay (NaBr):≥99.0 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Alkalinity (as Na₂CO₃):≤0.015 %
- ACS - pH of 5% Solution at 25°C:5.0 - 8.8
- ACS - Bromate (BrO₃):≤0.001 %
- ACS - Calcium (Ca):≤0.002 %
- ACS - Chloride (Cl):≤0.2 %
- ACS - Sulfate (SO₄):≤0.002 %
- ACS - Barium (Ba):≤0.002 %
- ACS - Magnesium (Mg):≤0.001 %
- ACS - Potassium (K):≤0.1 %
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together