Order Entry
Switzerland
ContactUsLinkComponent
Potassium sulphate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Potassium sulphate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 7140-20
CAS Number:  
undefined
Potassium sulphate ≥99.0%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # 7140-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • ACS - Assay (K₂SO₄):
    ≥99.0 %
  • Solubility:
    Passes Test
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • ACS - Calcium (Ca):
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Nitrogen Compounds (as N):
    ≤5 ppm
  • ACS - Sodium (Na):
    ≤0.02 %
  • ACS - pH of 5% Solution at 25°C:
    5.5 - 8.5

Specification Test Results

About this item

 :
 :174.26 g/mol
Merck Index:13,07759
Boiling Pt:1698 °C (1013 hPa)
Melting Pt:1069 °C
Density:2.66 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-915-5
CAS Number:7778-80-5
Formula:K₂SO₄
MDL Number:MFCD00011388