Order Entry
Switzerland
ContactUsLinkComponent
Potassium iodide ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Potassium iodide ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Potassium iodide ≥99.0%, granular, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (KI):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • Loss on Drying at 150°C:
    ≤0.2 %
  • pH of 5% Solution at 25°C:
    6.0 - 9.2
  • Chloride and Bromide (as Cl):
    ≤0.01 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.005 %
  • Barium (Ba):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.002 %
  • Magnesium (Mg):
    ≤0.001 %
  • Sodium (Na):
    ≤0.005 %
  • Iodate (IO₃):
    ≤3 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤2.0 ppm

Specification Test Results

Related Information

Frequently Bought Together