Order Entry
Switzerland
ContactUsLinkComponent
 
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Catalog # 3149-09
Supplier:  Avantor
CAS Number:  
undefined
Potassium hydroxide 45% in aqueous solution, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Catalog # 3149-09
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    600 lb
  • Seq:
    7350

Specifications

Specification Test Results

  • Assay (KOH):
    45.0 - 46.5 %
  • Color (APHA):
    ≤10
  • Appearance:
    Passes Test
  • Density (g/mL) at 25°C:
    1.45 - 1.47
  • Identification:
    Passes Test
  • Insoluble Matter:
    ≤30 ppm
  • Potassium Carbonate (K₂CO₃):
    ≤1000 ppm
  • Ammonium Hydroxide Precipitate:
    ≤30 ppm
  • Chloride (Cl):
    ≤30 ppm
  • Nitrogen Compounds (as N):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • ACS - Sulfate (SO₄):
    ≤5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤5.000 ppm
  • Trace Impurities - Chromium (Cr):
    ≤5.0 ppm
  • Trace Impurities - Copper (Cu):
    ≤0.050 ppm
  • Heavy Metals (as Ag):
    ≤3 ppm
  • Trace Impurities - Iron (Fe):
    ≤1 ppm
  • Trace Impurities - Lead (Pb):
    ≤2.000 ppm
  • Trace Impurities - Mercury (Hg):
    ≤0.10 ppm
  • Trace Impurities - Nickel (Ni):
    ≤1.000 ppm
  • Trace Impurities - Sodium (Na):
    ≤200.000 ppm
  • Trace Impurities - Zinc (Zn):
    ≤5.000 ppm

Specification Test Results

About this item

 :
 :56.11 g/mol
UN:1814
ADR:8,II
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553