Order Entry
Switzerland
ContactUsLinkComponent
Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®
Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Potassium chloride 99.0-100.5% (dried basis), crystals USP, FCC, J.T.Baker®
Supplier:  Avantor
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • Food GMP Manufactured Product:
  • Meets F.C.C. Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Food Chemical:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (KCl) (dried basis):
    99.0 - 100.5 %
  • USP - Acidity or Alkalinity:
    Passes Test
  • USP - Calcium and Magnesium:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Bromide (Br):
    ≤0.1 %
  • USP - Iodides (I):
    ≤0.005 %
  • USP - Loss on Drying:
    ≤1.0 %
  • USP - Sodium (Na):
    Passes Test
  • FCC - Assay (KCl) (dried basis):
    99.0 - 100.5 %
  • FCC - Acidity or Alkalinity:
    Passes Test
  • FCC - Heavy Metals (as Pb):
    ≤5 ppm
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Iodide and/or Bromide:
    Passes Test
  • FCC - Loss on Drying:
    ≤1.0 %
  • FCC - Sodium (Na):
    Passes Test
  • Appearance (White crystals):
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together