Order Entry
Switzerland
ContactUsLinkComponent
Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®
Irritant
Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Orthophosphoric acid 10% NF, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Alkali Phosphates:
    Passes Test
  • NF - Identification:
    Passes Test
  • NF - Nitrate (NO₃):
    Passes Test
  • NF - Phosphorous or Hypophosphorous Acid:
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Assay (as H₃PO₄), g/100 mL:
    9.5 - 10.5
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤ 6 ppm
  • EP/BP - Phosphorous or Hypophosphorous Acid:
    Passes Test
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 6 ppm
  • EP/BP - Substances Precipitated with Ammonia:
    Passes Test
  • EP/BP - Sulfate (SO₄):
    ≤ 10 ppm
  • EP/BP - Assay (H₃PO₄):
    9.5 - 10.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together