Order Entry
Switzerland
ContactUsLinkComponent
 
Orthophosphoric acid ≥85%, Finyte® for microelectronic, J.T.Baker®
  5854-05
 :  Avantor
 :  
undefined
Orthophosphoric acid ≥85%, Finyte® for microelectronic, J.T.Baker®
  5854-05
 :  Avantor
 :  

 

  • Pk:
    14 lb
  • Seq:
    7104

 

 

  • For Microelectronic Use:
    85.0 - 87.0 %
  • Store at temperatures above 22°C (72°F):
    = 10
  • Assay (H₃PO₄) (by acidimetry):
    85.0 - 87.0 %
  • Color (APHA):
    ≤10
  • Volatile Acids (µeq/g):
    ≤0.16
  • Chloride (Cl):
    ≤0.7 ppm
  • Nitrate (NO₃):
    ≤1.5 ppm
  • Sulfate (SO₄):
    ≤8 ppm
  • Trace Impurities - Aluminum (Al):
    ≤400.0 ppb
  • Trace Impurities - Antimony (Sb):
    ≤6000.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤50.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤450.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤200.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤50.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤50 ppb
  • Trace Impurities - Gold (Au):
    ≤150.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤700.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤100 ppb
  • Trace Impurities - Lithium (Li):
    ≤100.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤100.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤100.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤200.0 ppb
  • Trace Impurities - Potassium (K):
    ≤200.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤500.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤100.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤300.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤300.0 ppb
  • Particle Count at point of fill - 0.5 µm and greater:
    ≤50 par/ml
  • Particle Count at point of fill - 1.0 µm and greater:
    ≤10 par/ml

 

 

 :
 :98 g/mol
 :1805
 :8,III
 :7664-38-2
 :H₃PO₄
 :MFCD00011340