Order Entry
Switzerland
ContactUsLinkComponent
Petroleum spirit, 40…60 °C, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…60 °C, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 5280.2500
Supplier:  Avantor
CAS Number:  
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…60 °C, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 5280.2500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Acidity:
    max. 0.0005 meq/g
  • Boiling Range (initial to dry point):
    40 - 60 °C
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Water (KF):
    max. 0.02 %

Specification Test Results

About this item

 :
UN:1268
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849