Order Entry
Switzerland
ContactUsLinkComponent
N,N-Dimethylacetamide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
N,N-Dimethylacetamide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
Catalog # BAKR9755.1000
Supplier:  Avantor
CAS Number:  
undefined
N,N-Dimethylacetamide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
Catalog # BAKR9755.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.9 %
  • Acidity (meq/g):
    max. 0.0005
  • Evaporation Residue:
    max. 0.0005 %
  • GC-HS Suitability:
    passes test
  • Water (KF):
    max. 0.05 %

Specification Test Results

About this item

 :
 :87.12 g/mol
UN:2810
ADR:6.1,III
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686