

Dichloromethane ≥99.0%, GenAR® NF, Macron Fine Chemicals™
Catalog # 4874-26
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Steel drum
- Pk:200 L
- Seq:5549
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:= 99.0 %
- NF - Assay:≥99.0 %
- NF - Identification:Passes Test
- NF - Free Chlorine (Cl₂):Passes Test
- NF - Hydrogen Chloride (HCl):≤0.001 %
- NF - Methanol:≤600 ppm
- NF - Nonvolatile Residue:≤0.002 %
- NF - Specific Gravity at 25°C:1.318 - 1.322
- NF - Water:≤0.02 %
- 1,1-dichloroethene:—
- EP - Appearance:Passes Test
- EP - Refractive Index:1.423 - 1.425 Degree
- EP - Identification B:Passes Test
- EP - Identification C:Passes Test
- EP - Free Chlorine:Passes Test
- EP - Acidity:Passes Test
- EP - Residue on Evaporation:≤20 ppm
- EP - Relative Density @ 20° C:1.320 - 1.332
- EP - Water (KF):≤0.02 %
- EP - Ethanol, v/v:≤2.0 %
- EP - 2-methylbut-2-ene, v/v:≤300 ppm
- EP - Carbon tetrachloride, v/v:≤10 ppm
- EP - Chloroform, v/v:≤50 %
- EP - Total other impurities v/v:≤0.1 %
Specification Test Results
About this item
:
:84.93 g/mol
Density:1.32 g/cm³
UN:1593
ADR:6.1,III
:GHS07
:GHS08
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881