


4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), CMOS for microelectronic, J.T.Baker®
Catalog # 9405-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:8 pt US
- Seq:8026
Specifications
Specification Test Results
- For Microelectronic Use:Passes Test
- Appearance:Passes Test
- Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water):≥98.5 %
- Color (APHA):≤15
- Density (g/mL) at 25°C:0.794 - 0.798
- Residue after Evaporation:≤0.0050 %
- Acidity (as CH₃COOH):≤0.01 %
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
- Particle Count - 0.5 µm and greater:≤130 par/ml
- Particle Count - 1.0 µm and greater:≤10 par/ml
- Boiling Range (Initial):≥114.0 °C
- Boiling Range (Dry Point):≤117.0 °C
Specification Test Results
About this item
:
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938