


Hydrofluoric acid 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Référence 9387.0500
Fournisseur: Avantor
Numéro CAS:
Spécifications
- Pk:500 mL
- Seq:2725
Spécifications
Résultats des tests de spécification
- Trace Impurities - Yttrium (Y):≤0.1 ppb
- Trace Impurities - Zinc (Zn):≤1 ppb
- Trace Impurities - Zirconium (Zr):≤0.1 ppb
- Samarium (Sm):max. 0.1
- For Laboratory,Research,or Manufacturing Use:47 - 51 %
- Color (APHA):≤10
- Assay (HF)(w/w):47 - 51 %
- Chloride (Cl):≤4 ppm
- Fluosilicic Acid (SiF₆):≤20 ppm
- Total Phosphorus (P):≤0.05 ppm
- Total Sulphur (S):≤0.1 ppm
- Trace Impurities - Aluminum (Al):≤1 ppb
- Trace Impurities - Antimony (Sb):≤0.2 ppb
- Trace Impurities - Arsenic (As):≤0.5 ppb
- Trace Impurities - Barium (Ba):≤0.1 ppb
- Trace Impurities - Beryllium (Be):≤0.1 ppb
- Trace Impurities - Bismuth (Bi):≤0.1 ppb
- Trace Impurities - Boron (B):≤1 ppb
- Trace Impurities - Cadmium (Cd):≤0.1 ppb
- Trace Impurities - Calcium (Ca):≤1 ppb
- Trace Impurities - Cerium (Ce):≤0.1 ppb
- Trace Impurities - Cesium (Cs):≤0.1 ppb
- Trace Impurities - Chromium (Cr):≤1 ppb
- Trace Impurities - Cobalt (Co):≤0.1 ppb
- Trace Impurities - Copper (Cu):≤0.5 ppb
- Trace Impurities - Dysprosium (Dy):≤0.1 ppb
- Trace Impurities - Erbium (Er):≤0.1 ppb
- Trace Impurities - Europium (Eu):≤0.1 ppb
- Trace Impurities - Gadolinium (Gd):≤0.1 ppb
- Trace Impurities - Gallium (Ga):≤0.1 ppb
- Trace Impurities - Germanium (Ge):≤0.1 ppb
- Trace Impurities - Gold (Au):≤0.2 ppb
- Trace Impurities - Hafnium (Hf):≤0.1 ppb
- Trace Impurities - Holmium (Ho):≤0.1 ppb
- Trace Impurities - Indium (In):≤0.1 ppb
- Trace Impurities - Iron (Fe):≤1 ppb
- Trace Impurities - Lanthanum (La):≤0.1 ppb
- Trace Impurities - Lead (Pb):≤0.1 ppb
- Trace Impurities - Lithium (Li):≤0.1 ppb
- Trace Impurities - Lutetium (Lu):≤0.1 ppb
- Trace Impurities - Magnesium (Mg):≤1 ppb
- Trace Impurities - Manganese (Mn):≤0.1 ppb
- Trace Impurities - Mercury (Hg):≤1 ppb
- Trace Impurities - Molybdenum (Mo):≤0.1 ppb
- Trace Impurities - Neodymium (Nd):≤0.1 ppb
- Trace Impurities - Nickel (Ni):≤0.5 ppb
- Trace Impurities - Niobium (Nb):≤0.1 ppb
- Trace Impurities - Palladium (Pd):max. 0.1
- Trace Impurities - Platinum (Pt):max. 1
- Trace Impurities - Potassium (K):≤1 ppb
- Trace Impurities - Praseodymium (Pr):≤0.1 ppb
- Trace Impurities - Rhenium (Re):≤0.1 ppb
- Trace Impurities - Rhodium (Rh):≤0.1 ppb
- Trace Impurities - Rubidium (Rb):≤0.1 ppb
- Trace Impurities - Ruthenium (Ru):≤0.1 ppb
- Trace Impurities - Samarium (Sm):≤0.1 ppb
- Trace Impurities - Scandium (Sc):≤0.1 ppb
- Trace Impurities - Selenium (Se):≤1 ppb
- Trace Impurities - Silver (Ag):≤0.5 ppb
- Trace Impurities - Sodium (Na):≤1 ppb
- Trace Impurities - Strontium (Sr):≤0.1 ppb
- Trace Impurities - Tantalum (Ta):max. 0.1
- Trace Impurities - Tellurium (Te):≤0.1 ppb
- Trace Impurities - Terbium (Tb):≤0.1 ppb
- Trace Impurities - Thallium (Tl):≤0.1 ppb
- Trace Impurities - Thorium (Th):≤0.1 ppb
- Trace Impurities - Thulium (Tm):≤0.1 ppb
- Trace Impurities - Tin (Sn):≤0.5 ppb
- Trace Impurities - Titanium (Ti):≤1 ppb
- Trace Impurities - Tungsten (W):≤0.5 ppb
- Trace Impurities - Uranium (U):≤0.1 ppb
- Trace Impurities - Vanadium (V):≤0.1 ppb
- Trace Impurities - Ytterbium (Yb):≤0.1 ppb
Résultats des tests de spécification
À propos de cet article
:
:20.01 g/mol
ADR:8,I
:Management of Change (MOC) category = N
EINECS:231-634-8
UN:1790
:GHS06
:GHS05
:GHS08
Température de stockage:Ambient
Numéro CAS:7664-39-3
Formule:HF
Numéro MDL:MFCD00011346