Order Entry
Switzerland
ContactUsLinkComponent
 
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, ACS, J.T.Baker®
Catalog # 2262-20
Supplier:  Avantor
CAS Number:  
undefined
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular USP, ACS, J.T.Baker®
Catalog # 2262-20
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE drum, open head
  • Pk:
    100 kg
  • Seq:
    4553

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C: Water (H₂O):
    ≤1.0 %
  • USP - Assay (Anhydrous basis):
    97.5 - 102.0 %
  • USP - Related substances: Maltose and isomaltose:
    ≤0.4 %
  • USP - Dextrin:
    Passes Test
  • USP - Related substances: Maltotriose:
    ≤0.2 %
  • USP - Related substances: Fructose:
    ≤0.15 %
  • USP - Related substances: Unspecified impurities, each:
    ≤0.10 %
  • USP - Related substances: Total Impurities:
    ≤0.5 %
  • USP - Color and Clarity of Solution:
    Passes Test
  • USP - Soluble Starch, Sulfites:
    ≤15 ppm
  • USP - Conductivity (uS cm-1):
    ≤20 µS/cm
  • Chloride (Cl):
    ≤0.01 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Insoluble Matter:
    ≤0.005 %
  • Loss on Drying at 105°C:
    ≤0.2 %
  • Residue after Ignition:
    ≤0.015 %
  • Solution in Water (50%)(APHA):
    ≤20
  • Specific Rotation [Å]²⁵ ^D (+):
    52.5 - 53.0 Degree
  • Starch:
    Passes Test
  • Sulfate and Sulfite (as SO₄):
    ≤0.005 %
  • Titrable Acid (meq/g):
    ≤0.002
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :180.16 g/mol
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912