Order Entry
Switzerland
ContactUsLinkComponent
Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Irritant
Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
    99.5 - 100.5%
  • Meets U.S.P Requirements:
    Passes Test
  • Preserve in Tight Containers:
    Passes Test
  • CAUTION: For Manufacturing, processing or repackaging:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.1%
  • USP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):
    99.5 - 100.5 %
  • USP - Clarity of Solution:
    Passes Test
  • USP - Color of Solution:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Oxalic Acid:
    ≤0.036 %
  • USP - Sulfate (SO₄):
    ≤0.015 %
  • USP - Readily Carbonizable Substances:
    Passes Test
  • USP - Identification C: Water (H₂O):
    ≤1.0 %

Specification Test Results

Related Information

Frequently Bought Together