

Chloroform ≥99.8% (by GC, corrected for water content), purified, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:05-2114614906-45
:GHS06
:GHS08
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.8 %
- Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water):≥99.8 %
- Specific Gravity at 25°/25°C:1.476 - 1.480
- Nonvolatile Residue (w/v):≤0.002 %
- Free Chlorine:Passes Test
- Readily Carbonizable Substances:Passes Test
- Chlorinated Decomposition Products and Chloride:Passes Test
- Acid and Phosgene:Passes Test
- Aldehydes and Ketones (as HCHO):Passes Test
Specification Test Results
Frequently Bought Together