

Chloroform ≥99.8% (by GC), AR® ACS, Macron Fine Chemicals™
Catalog # 4440-24
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Steel drum, phenolic lined
- Pk:650 lb
- Seq:7359
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.8%
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance (clear, colorless liquid):Passes Test
- ACS - Assay (CHCl₃) (by GC):≥99.8 %
- ACS - Acetone, Aldehydes (as (CH₃)₂CO):Passes Test
- ACS - Acid and Chloride:Passes Test
- ACS - Color (APHA):≤10
- ACS - Free Chlorine:Passes Test
- ACS - Lead (Pb):≤0.050 ppm
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Suitability for Use in Dithizone Tests:Passes Test
Specification Test Results
About this item
:
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-663-8
UN:1888
ADR:6.1,III
REACH:05-2114614906-45
:GHS08
:GHS07
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826