
Calcium chloride dihydrate 99.0-107.0%, CalPlus™ USP, Multi-Compendial, J.T.Baker®
: Avantor
:10035-04-8
:
:CaCl₂·2H₂O
:MFCD00149613
:147.01 g/mol
:13,01660
:1600 °C (1013 hPa)
:176 °C
:1.85 g/cm³ (20 °C)
:Management of Change (MOC) category = R
:233-140-8
:GHS07
:Ambient
- Meets F.C.C. Requirements:—
- Meets U.S.P Requirements:—
- GMP Manufactured Product:—
- Meets J.P. Chemical Specifications:—
- Meets E.P. Chemical Specifications:—
- Food GMP Manufactured Product:—
- Meets B.P. Chemical Specifications:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Food Chemical:—
- Bulk Pharmaceutical Chemical:—
- USP - Assay (CaCl₂·2H₂O):99.0 - 107.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH of 5% Solution at 25°C:4.5 - 9.2
- USP - Iron, Aluminum, and Phosphate:Passes Test
- USP - Magnesium and Alkali Salts:≤ 1.0 %
- USP - Aluminum (Al)(µg/g):≤ 1
- Endotoxin Concentration (EU/g):≤ 10
- FCC - Assay (CaCl₂·2H₂O):99.0 - 107.0 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Arsenic (As):≤ 3 mg/kg
- FCC - Magnesium and Alkali Salts:≤ 1.0 %
- FCC - Lead (Pb):≤ 5 mg/kg
- FCC - Fluoride (F):≤ 0.004 %
- Appearance (White crystals or granules):Passes Test
- EP/BP - Assay (CaCl₂·2H₂O):97.0 - 103.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP - Aluminum (Al):≤ 1 ppm
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Magnesium and Alkali Metals:≤ 0.5 %
- JP - Assay (CaCl₂·2H₂O):96.7 - 103.3 %
- JP - Identification:Passes Test
- JP - pH (1 in 20):4.5 - 9.2
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Clarity and Color of Solution:Passes Test
- JP - Sulfate (SO₄):≤ 0.024 %
- JP - Hypochlorite:Passes Test
- JP - Iron, Aluminum or Phosphate:Passes Test
- JP - Barium (Ba):Passes Test
- JP - Arsenic (As):≤ 2 ppm
- No Class 1,2,3 or other solvents are used or produced in the:≤ 2 ppm
Frequently Bought Together